Anti-AFAP1L1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB25744
ABNOPAB25744EA 585 EUR
ABNOPAB25744
Anti-AFAP1L1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of AFAP1L1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 5 ?g/ml

Type: Primary
Antigen: AFAP1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR