Anti-CEP162 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB22408
ABNOPAB22408EA 523 EUR
ABNOPAB22408
Anti-CEP162 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant KIAA1009.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CEP162
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR