Anti-PI4K2B Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB20260
ABNOPAB20260EA 523 EUR
ABNOPAB20260
Anti-PI4K2B Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant PI4K2B.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PI4K2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

TEKNISET TIEDOT

Antigeenin symboli PI4K2B
Environmentally Preferable
Antigeenin nimi Phosphatidylinositol 4-kinase Type 2 Beta
Antigeenin synonyymit PIK42B|PI4KIIB
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Unconjugated
Reaktiivisuus Human
Isäntä Rabbit
Geeni ID 55300
Isotyyppi IgG
Western blot Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Immunogeeni Recombinant protein corresponding to amino acids of human PI4K2B.
Sekvenssi LILPYISDMNFVQDLCEDLYELFKTDKGFDKATFESQMSVMRGQILNLTQALRDGKSPFQLVQIPCVIVERSQGGSQGRIVHLSNSFTQTVNCRKPF
Muoto Liquid
Puhdistus Antigen affinity purification
Ristiadsorptio No
Säilytyspuskuri PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
Säilytyslämpötila Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR