Anti-ANXA6 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB19522
ABNOPAB19522EA 523 EUR
ABNOPAB19522
Anti-ANXA6 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of ANXA6.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: ANXA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR