Anti-AQP9 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB18840
ABNOPAB18840EA 423 EUR
ABNOPAB18840
Anti-AQP9 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of AQP9.

Recommended Dilutions: ELISA: 1:80000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR