Anti-PLA2G4A Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB18118
ABNOPAB18118EA 423 EUR
ABNOPAB18118
Anti-PLA2G4A Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of PLA2G4A.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:10000

Type: Primary
Antigen: PLA2G4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR