Anti-SYT6 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB17103
ABNOPAB17103EA 423 EUR
ABNOPAB17103
Anti-SYT6 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic phosphopeptide of SYT6.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SYT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR