Anti-Borrelia burgdorferi Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB16653
ABNOPAB16653EA 912 EUR
ABNOPAB16653
Anti-Borrelia burgdorferi Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against native Borrelia burgdorferi.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: Borrelia burgdorferi
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR