Anti-NUDC Goat Polyclonal Antibody

Toimittaja: Abnova
PAB16636
ABNOPAB16636EA 523 EUR
ABNOPAB16636
Anti-NUDC Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody raised against synthetic peptide of NUDC.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: NUDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR