Anti-NDR1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB16023
ABNOPAB16023EA 648 EUR
ABNOPAB16023
Anti-NDR1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of NDR1.

Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: NDR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Thale cress
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR