Anti-DICER1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB15829
ABNOPAB15829EA 648 EUR
ABNOPAB15829
Anti-DICER1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against partial recombinant Dicer1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: DICER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR