Anti-BLNK Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB13541
ABNOPAB13541EA 402 EUR
ABNOPAB13541
Anti-BLNK Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against partial recombinant BLNK.

Type: Primary
Antigen: BLNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR