Anti-MAP3K5 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB13430
ABNOPAB13430EA 585 EUR
ABNOPAB13430
Anti-MAP3K5 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K5.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR