Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB13412
ABNOPAB13412EA 585 EUR
ABNOPAB13412
Anti-West Nile Virus Envelope Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Envelope.

Recommended Dilutions: ELISA: 1 ?g/ml

Type: Primary
Antigen: West Nile Virus Envelope
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR