Anti-TARDBP Goat Polyclonal Antibody

Toimittaja: Abnova
PAB11464
ABNOPAB11464EA 523 EUR
ABNOPAB11464
Anti-TARDBP Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody raised against synthetic peptide of TARDBP.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: TARDBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR