Anti-S1PR4 Polyclonal Antibody

Toimittaja: Abnova
PAB10364
ABNOPAB10364EA 912 EUR
ABNOPAB10364
Anti-S1PR4 Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of S1PR4.

Recommended Dilutions: Western Blot: 5 to 10 ?g/ml

Type: Primary
Antigen: S1PR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR