Anti-TLX1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB0355
ABNOPAB0355EA 523 EUR
ABNOPAB0355
Anti-TLX1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of TLX1.

Type: Primary
Antigen: TLX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR