Anti-AMBP Rabbit Polyclonal Antibody

Toimittaja: Abnova
PAB0125
ABNOPAB0125EA 648 EUR
ABNOPAB0125
Anti-AMBP Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against Ambp.

Recommended Dilutions: Western Blot: 1:2000; Radio Immunoassay: 1:6000; ELISA: 1:5000; Immunohistochemistry: 1:200

Type: Primary
Antigen: AMBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR