Anti-IgG Mouse Monoclonal Antibody [clone: EM-07] (FITC (Fluorescein Isothiocyanate))

Toimittaja: Abnova
MAB9827
ABNOMAB9827EA 402 EUR
ABNOMAB9827
Anti-IgG Mouse Monoclonal Antibody [clone: EM-07] (FITC (Fluorescein Isothiocyanate))
Vasta-aineet
Mouse monoclonal antibody raised against IgG (Fc).

Recommended Dilutions: Immunofluorescence: 2-5 ?g/ml

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: EM-07
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(c)
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR