Anti-MOMP Mouse Monoclonal Antibody [clone: CL12-671.11.23]

Toimittaja: Abnova
MAB6152
ABNOMAB6152EA 402 EUR
ABNOMAB6152
Anti-MOMP Mouse Monoclonal Antibody [clone: CL12-671.11.23]
Vasta-aineet
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.

Type: Primary
Antigen: MOMP
Clonality: Monoclonal
Clone: CL12-671.11.23
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: C. trachomatis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR