Anti-ITGA4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PS/2]

Toimittaja: Abnova
MAB6099
ABNOMAB6099EA 402 EUR
ABNOMAB6099
Anti-ITGA4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: PS/2]
Vasta-aineet
Rat monoclonal antibody raised against native Itga4.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: PS/2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR