Anti-EDN1 Mouse Monoclonal Antibody [clone: 403]

Toimittaja: Abnova
MAB4685
ABNOMAB4685EA 1690 EUR
ABNOMAB4685
Anti-EDN1 Mouse Monoclonal Antibody [clone: 403]
Vasta-aineet
Mouse monoclonal antibody raised against EDN1.

Type: Primary
Antigen: EDN1
Clonality: Monoclonal
Clone: 403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR