Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11[APG]]

Toimittaja: Abnova
MAB3835
ABNOMAB3835EA 402 EUR
ABNOMAB3835
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11[APG]]
Vasta-aineet
Mouse monoclonal antibody raised against native CD34.

Recommended Dilutions: Flow Cytometry: 2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml; Western Blot: 2 ?g/ml

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11[APG]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR