Anti-TNFSF4 Rat Monoclonal Antibody [clone: MPG34]

Toimittaja: Abnova
MAB3725
ABNOMAB3725EA 1170 EUR
ABNOMAB3725
Anti-TNFSF4 Rat Monoclonal Antibody [clone: MPG34]
Vasta-aineet
Rat monoclonal antibody raised against Tnfsf4.

Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: MPG34
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR