Anti-TNN1 Mouse Monoclonal Antibody [clone: T20]

Toimittaja: Abnova
MAB1529
ABNOMAB1529EA 523 EUR
ABNOMAB1529
Anti-TNN1 Mouse Monoclonal Antibody [clone: T20]
Vasta-aineet
Mouse monoclonal antibody raised against native TNN.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml; Immunohistochemistry (Frozen sections): 2-4 ?g/ml

Type: Primary
Antigen: TNN1
Clonality: Monoclonal
Clone: T20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR