Anti-MLH1 Mouse Monoclonal Antibody [clone: 4C9C7]

Toimittaja: Abnova
MAB15272
ABNOMAB15272EA 423 EUR
ABNOMAB15272
Anti-MLH1 Mouse Monoclonal Antibody [clone: 4C9C7]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant human MLH1.

Type: Primary
Antigen: MLH1
Clonality: Monoclonal
Clone: 4C9C7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR