Anti-DMD Mouse Monoclonal Antibody [clone: DYS-48]

Toimittaja: Abnova
MAB1453
ABNOMAB1453EA 523 EUR
ABNOMAB1453
Anti-DMD Mouse Monoclonal Antibody [clone: DYS-48]
Vasta-aineet
Mouse monoclonal antibody raised against full length recombinant DMD.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: DMD
Clonality: Monoclonal
Clone: DYS-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR