Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]

Toimittaja: Abnova
MAB1437
ABNOMAB1437EA 523 EUR
ABNOMAB1437
Anti-CDC6 Mouse Monoclonal Antibody [clone: IMD-6]
Vasta-aineet
Mouse monoclonal antibody raised against full length recombinant CDC6.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CDC6
Clonality: Monoclonal
Clone: IMD-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR