Anti-c-Myc tag Monoclonal Antibody

Toimittaja: Abnova
MAB1296
ABNOMAB1296EA 402 EUR
ABNOMAB1296
Anti-c-Myc tag Monoclonal Antibody
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptides of c-Myc tag.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: c-Myc tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR