Anti-BRAF Mouse Monoclonal Antibody [clone: 1H12]

Toimittaja: Abnova
MAB10422
ABNOMAB10422EA 648 EUR
ABNOMAB10422
Anti-BRAF Mouse Monoclonal Antibody [clone: 1H12]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant BRAF.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: 1H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR