Cell proliferation assay kit, WST-1
Toimittaja: Abnova
KA1384
ABNOKA1384EA 140 EUR
ABNOKA1384
Cell proliferation assay kit, WST-1
Bio-assayt Solujen testausanalyysit
WST-1 cell proliferation assay kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.
  • Application: functional study
  • Store the kit at –20 °C
Varoitus: For research use only (RUO).