Anti-WNT5B Mouse Polyclonal Antibody

Toimittaja: Abnova
H00081029-B01P
ABNOH00081029-B01PEA 397 EUR
ABNOH00081029-B01P
Anti-WNT5B Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: WNT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR