Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 3E5]

Toimittaja: Abnova
H00030008-M33
ABNOH00030008-M33EA 397 EUR
ABNOH00030008-M33
Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 3E5]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant EFEMP2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: EFEMP2
Clonality: Monoclonal
Clone: 3E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR