Anti-RHOD Mouse Monoclonal Antibody [clone: 1E7]

Toimittaja: Abnova
H00029984-M12
ABNOH00029984-M12EA 397 EUR
ABNOH00029984-M12
Anti-RHOD Mouse Monoclonal Antibody [clone: 1E7]
Vasta-aineet
Type: Primary
Antigen: RHOD
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR