Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 3F10]

Toimittaja: Abnova
H00023428-M01
ABNOH00023428-M01EA 397 EUR
ABNOH00023428-M01
Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 3F10]
Vasta-aineet
Type: Primary
Antigen: SLC7A8
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR