Anti-SKIV2L Mouse Polyclonal Antibody

Toimittaja: Abnova
H00006499-A01
ABNOH00006499-A01EA 260 EUR
ABNOH00006499-A01
Anti-SKIV2L Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SKIV2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR