Anti-LRP1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
H00004035-D01P
ABNOH00004035-D01PEA 364 EUR
ABNOH00004035-D01P
Anti-LRP1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: LRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR