Anti-DDX11 Mouse Monoclonal Antibody [clone: 3C1]

Toimittaja: Abnova
H00001663-M03
ABNOH00001663-M03EA 397 EUR
ABNOH00001663-M03
Anti-DDX11 Mouse Monoclonal Antibody [clone: 3C1]
Vasta-aineet
Type: Primary
Antigen: DDX11
Clonality: Monoclonal
Clone: 3C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR