Anti-DAD1 Mouse Polyclonal Antibody

Toimittaja: Abnova
H00001603-A01
ABNOH00001603-A01EA 260 EUR
ABNOH00001603-A01
Anti-DAD1 Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: DAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR