Anti-CKMT2 Mouse Polyclonal Antibody

Toimittaja: Abnova
H00001160-B01P
ABNOH00001160-B01PEA 397 EUR
ABNOH00001160-B01P
Anti-CKMT2 Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human CKMT2 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CKMT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR