Anti-IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon) Rabbit Polyclonal Antibody [clone: 421CT8.1.1]

Toimittaja: Abgent
AP8138B
ABGEAP8138BEA 354 EUR
ABGEAP8138B
Anti-IKBKE (inhibitor of kappa light Polypeptide gene enhancer in B-cells, kinase epsilon) Rabbit Polyclonal Antibody [clone: 421CT8.1.1]
Vasta-aineet
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR