Anti-PRKG2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP8001A
ABGEAP8001AEA 354 EUR
ABGEAP8001A
Anti-PRKG2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PRKG2 (protein kinase, cGMP-dependent, type II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR