Anti-MAP2K1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP7960A
ABGEAP7960AEA 354 EUR
ABGEAP7960A
Anti-MAP2K1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR