Anti-RAB5A Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP6974B
ABGEAP6974BEA 354 EUR
ABGEAP6974B
Anti-RAB5A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RAB5A (RAB5A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR