Anti-ESRP1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP5940A
ABGEAP5940AEA 354 EUR
ABGEAP5940A
Anti-ESRP1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ESRP1 (epithelial splicing regulatory protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR