Anti-FFAR3 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP5222B
ABGEAP5222BEA 354 EUR
ABGEAP5222B
Anti-FFAR3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: FFAR3 (free fatty acid receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR