Anti-AKT3 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP3468A
ABGEAP3468AEA 354 EUR
ABGEAP3468A
Anti-AKT3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR