Anti-MEF2C Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP3382A
ABGEAP3382AEA 354 EUR
ABGEAP3382A
Anti-MEF2C Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR