Anti-CRK Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP3304A
ABGEAP3304AEA 354 EUR
ABGEAP3304A
Anti-CRK Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CRK (v-crk avian sarcoma virus CT10 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR