Anti-RAD9A Rabbit Polyclonal Antibody
Toimittaja: Abgent
AP3224A
ABGEAP3224AEA 354 EUR
ABGEAP3224A
Anti-RAD9A Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human